siRNA Details
Gene Symbol: SCAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290229.1 | NP_001277158.1 | 8 | 961 | ||
NM_004866.5 | NP_004857.4 | 9 | 1039 | |||
NR_110885.1 | 8 | 879 | ||||
XM_011543727.2 | XP_011542029.1 | 8 | 937 | |||
GenBank mRNA | AF005037.1 | - | 9 | 913 | ||
AF038966.1 | - | 9 | 960 | |||
AK001541.1 | - | 3 | 267 | |||
AK225172.1 | - | 9 | 928 | |||
AK290225.1 | - | 9 | 889 | |||
AK298146.1 | - | 7 | 672 | |||
AK304452.1 | - | 8 | 853 | |||
BC009787.1 | - | 8 | 771 | |||
BC015065.1 | - | 9 | 932 | 4105068 | ||
BC016355.1 | - | 3 | 297 | |||
BC034048.1 | - | 9 | 893 | 4814951 | ||
CR542286.1 | - | 9 | 870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
