siRNA Details
Gene Symbol: SEMA4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324029.1 | NP_001310958.1 | 13 | 1993 | ||
NM_001324030.1 | NP_001310959.1 | 14 | 2175 | |||
NM_001324031.1 | NP_001310960.1 | 15 | 2361 | |||
NM_001324032.1 | NP_001310961.1 | 14 | 2438 | |||
NM_001324033.1 | NP_001310962.1 | 15 | 2447 | |||
NM_001324034.1 | NP_001310963.1 | 14 | 2449 | |||
NM_020210.3 | NP_064595.2 | 15 | 2453 | |||
NM_198925.2 | NP_945119.1 | 14 | 2419 | |||
GenBank mRNA | AB051532.1 | - | 14 | 2407 | ||
AF258561.1 | - | 3 | 503 | |||
AK026108.1 | - | 14 | 2152 | |||
AK027083.1 | - | 1 | 403 | |||
AK289991.1 | - | 15 | 2422 | |||
AL390080.1 | - | 1 | 37 | |||
AL390081.1 | - | 10 | 1679 | |||
AL390082.1 | - | 1 | 109 | |||
AY358392.1 | - | 15 | 2410 | |||
AY445887.1 | - | 14 | 2430 | |||
BC010701.1 | - | 7 | 1251 | |||
BC017658.1 | - | 6 | 1033 | |||
BC019013.2 | - | 1 | 214 | |||
BC094849.1 | - | 7 | 1223 | |||
BC110853.1 | - | 14 | 2174 | 6020867 | ||
BC126438.1 | - | 14 | 2328 | 8992154 | ||
BC144141.1 | - | 15 | 2356 | |||
BX537671.1 | - | 11 | 2372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
