siRNA Details
Gene Symbol: SEMA4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324029.1 | NP_001310958.1 | 13 | 1906 | ||
NM_001324030.1 | NP_001310959.1 | 14 | 2088 | |||
NM_001324031.1 | NP_001310960.1 | 15 | 2274 | |||
NM_001324032.1 | NP_001310961.1 | 14 | 2351 | |||
NM_001324033.1 | NP_001310962.1 | 15 | 2360 | |||
NM_001324034.1 | NP_001310963.1 | 14 | 2362 | |||
NM_020210.3 | NP_064595.2 | 15 | 2366 | |||
NM_198925.2 | NP_945119.1 | 14 | 2332 | |||
GenBank mRNA | AB051532.1 | - | 14 | 2320 | ||
AF258561.1 | - | 3 | 416 | |||
AK026108.1 | - | 14 | 2065 | |||
AK026133.1 | - | 7 | 1236 | |||
AK027083.1 | - | 1 | 316 | |||
AK289991.1 | - | 15 | 2335 | |||
AL390081.1 | - | 10 | 1592 | |||
AL390082.1 | - | 1 | 22 | |||
AY358392.1 | - | 15 | 2323 | |||
AY445887.1 | - | 14 | 2343 | |||
BC010701.1 | - | 7 | 1164 | |||
BC017658.1 | - | 6 | 946 | |||
BC019013.2 | - | 1 | 127 | |||
BC094849.1 | - | 7 | 1136 | |||
BC110853.1 | - | 14 | 2087 | 6020867 | ||
BC126438.1 | - | 14 | 2241 | 8992154 | ||
BC144141.1 | - | 15 | 2269 | |||
BX537671.1 | - | 11 | 2285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
