siRNA Details
Gene Symbol: SEMA4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324029.1 | NP_001310958.1 | 9 | 1208 | ||
NM_001324030.1 | NP_001310959.1 | 10 | 1390 | |||
NM_001324031.1 | NP_001310960.1 | 11 | 1576 | |||
NM_001324032.1 | NP_001310961.1 | 10 | 1634 | |||
NM_001324033.1 | NP_001310962.1 | 11 | 1662 | |||
NM_001324034.1 | NP_001310963.1 | 10 | 1634 | |||
NM_020210.3 | NP_064595.2 | 11 | 1668 | |||
NM_198925.2 | NP_945119.1 | 10 | 1634 | |||
GenBank mRNA | AB051532.1 | - | 10 | 1622 | ||
AK026108.1 | - | 10 | 1367 | |||
AK026133.1 | - | 3 | 538 | |||
AK289991.1 | - | 11 | 1637 | |||
AL390081.1 | - | 6 | 894 | |||
AY358392.1 | - | 11 | 1625 | |||
AY445887.1 | - | 10 | 1645 | |||
BC010701.1 | - | 3 | 466 | |||
BC017658.1 | - | 2 | 248 | |||
BC094849.1 | - | 3 | 438 | |||
BC110853.1 | - | 10 | 1389 | 6020867 | ||
BC126438.1 | - | 10 | 1543 | 8992154 | ||
BC144141.1 | - | 11 | 1571 | |||
BX537671.1 | - | 7 | 1587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
