siRNA Details
Gene Symbol: C3orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178339.2 | NP_848029.2 | 6 | 1033 | ||
NR_126512.1 | 7 | 1019 | ||||
NR_126513.1 | 4 | 572 | ||||
NR_126514.1 | 4 | 669 | ||||
GenBank mRNA | AJ493599.1 | - | 7 | 1335 | ||
AJ493600.1 | - | 6 | 1349 | |||
AJ493601.1 | - | 4 | 888 | |||
AJ493602.1 | - | 4 | 985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
