siRNA Details
Gene Symbol: TCEB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016427.2 | NP_057511.2 | 1 | 728 | ||
GenBank mRNA | AB030834.1 | - | 1 | 489 | ||
AK314984.1 | - | 1 | 708 | |||
BC036022.1 | - | 1 | 732 | 5270406 | ||
BC103911.2 | - | 1 | 376 | 40006025 |
Gene Symbol: TCEB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145653.3 | NP_663628.2 | 1 | 611 | ||
GenBank mRNA | AB076840.1 | - | 1 | 375 |
Gene Symbol: TCEB3CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100817.1 | NP_001094287.1 | 1 | 375 | ||
GenBank mRNA | BC103912.1 | - | 1 | 371 | ||
BC103913.1 | - | 1 | 376 | 40006027 |
Gene Symbol: TCEB3CL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242907.1 | NP_001229836.1 | 1 | 611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
