siRNA Details
Gene Symbol: MXI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008541.1 | NP_001008541.1 | 5 | 2417 | ||
NM_005962.4 | NP_005953.4 | 6 | 2642 | |||
NM_130439.3 | NP_569157.2 | 6 | 2840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
