siRNA Details
Gene Symbol: MXI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008541.1 | NP_001008541.1 | 5 | 1736 | ||
NM_005962.4 | NP_005953.4 | 6 | 1961 | |||
NM_130439.3 | NP_569157.2 | 6 | 2159 | |||
GenBank mRNA | BC012907.1 | - | 5 | 1732 | 3882557 | |
BC035128.1 | - | 6 | 2160 | 5263647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
