siRNA Details
Gene Symbol: UXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004182.3 | NP_004173.1 | 5 | 406 | ||
NM_153477.2 | NP_705582.1 | 4 | 584 | |||
NR_045559.1 | 3 | 516 | ||||
NR_045560.1 | 5 | 406 | ||||
GenBank mRNA | AF083241.1 | - | 5 | 372 | ||
AF083242.1 | - | 4 | 522 | |||
AF092737.1 | - | 5 | 387 | |||
AK223320.1 | - | 5 | 360 | |||
AK312072.1 | - | 5 | 372 | |||
BC000720.1 | - | 4 | 341 | 3505415 | ||
BC008890.1 | - | 4 | 341 | 3959862 | ||
BM471483.1 | - | 3 | 443 | |||
BQ681820.1 | - | 1 | 80 | |||
EL949569.1 | - | 5 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
