siRNA Details
Gene Symbol: MAN1B1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027447.1 | 1 | 336 | |||
GenBank mRNA | AK023312.1 | - | 1 | 336 | ||
BC020858.1 | - | 1 | 190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
