siRNA Details
Gene Symbol: BRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018963.4 | NP_061836.2 | 16 | 1942 | ||
NM_033656.3 | NP_387505.1 | 16 | 1942 | |||
XM_011529611.1 | XP_011527913.1 | 16 | 1975 | |||
XM_011529612.2 | XP_011527914.1 | 9 | 838 | |||
XM_017028373.1 | XP_016883862.1 | 17 | 1747 | |||
XM_017028375.1 | XP_016883864.1 | 16 | 1975 | |||
GenBank mRNA | AB080586.1 | - | 16 | 1746 | ||
AB080587.1 | - | 16 | 1746 | |||
AB080588.1 | - | 8 | 781 | |||
AB080589.1 | - | 8 | 781 | |||
AB080591.1 | - | 8 | 781 | |||
AB080593.1 | - | 8 | 781 | |||
AJ238214.2 | - | 16 | 1746 | |||
AJ292465.1 | - | 16 | 1726 | |||
AJ292466.1 | - | 16 | 1726 | |||
AK125906.1 | - | 1 | 39 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
