siRNA Details
Gene Symbol: BRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018963.4 | NP_061836.2 | 41 | 5975 | ||
NM_033656.3 | NP_387505.1 | 41 | 5975 | |||
XM_011529611.1 | XP_011527913.1 | 40 | 5864 | |||
XM_011529612.2 | XP_011527914.1 | 34 | 4871 | |||
XM_011529613.1 | XP_011527915.1 | 19 | 2970 | |||
XM_017028373.1 | XP_016883862.1 | 42 | 5780 | |||
XM_017028374.1 | XP_016883863.1 | 17 | 4610 | |||
GenBank mRNA | AB080586.1 | - | 41 | 5779 | ||
AB080587.1 | - | 41 | 5779 | |||
AJ238214.2 | - | 41 | 5779 | |||
AJ292465.1 | - | 41 | 5759 | |||
AJ292466.1 | - | 41 | 5759 | |||
AK002177.1 | - | 2 | 748 | |||
AK125906.1 | - | 25 | 3927 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
