siRNA Details
Gene Symbol: LRRC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199340.3 | NP_955372.2 | 4 | 679 | ||
XM_017024593.1 | XP_016880082.1 | 2 | 508 | |||
XM_017024594.1 | XP_016880083.1 | 2 | 508 | |||
XM_017024595.1 | XP_016880084.1 | 2 | 508 | |||
XM_017024596.1 | XP_016880085.1 | 2 | 508 | |||
XM_017024597.1 | XP_016880086.1 | 2 | 508 | |||
XM_017024598.1 | XP_016880087.1 | 2 | 508 | |||
XM_017024600.1 | XP_016880089.1 | 2 | 508 | |||
GenBank mRNA | AB011135.2 | - | 3 | 544 | ||
BC043145.1 | - | 1 | 21 | 5296147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
