siRNA Details
Gene Symbol: MUC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002457.3 | NP_002448.3 | 12982 | |||
GenBank mRNA | AF257167.1 | - | 2 | 403 | ||
L21998.1 | - | 12982 | ||||
M22405.1 | - | 2 | 735 | |||
M94132.1 | - | 2 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
