siRNA Details
Gene Symbol: TTC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008237.2 | NP_001008238.1 | 3 | 513 | ||
GenBank mRNA | BC057850.1 | - | 3 | 513 | 4795916 | |
BC070381.1 | - | 3 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
