siRNA Details
Gene Symbol: COL25A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032518.2 | NP_115907.2 | 24, 25 | 1859 | ||
NM_198721.3 | NP_942014.1 | 24, 25 | 1859 | |||
NR_045756.2 | 21, 22 | 1233 | ||||
XM_011532333.2 | XP_011530635.1 | 24, 25 | 1952 | |||
XM_011532334.2 | XP_011530636.1 | 24, 25 | 1818 | |||
XM_011532335.2 | XP_011530637.1 | 24, 25 | 1647 | |||
XM_011532338.2 | XP_011530640.1 | 22, 23 | 2027 | |||
XM_011532355.2 | XP_011530657.1 | 23, 24 | 5731 | |||
XM_011532356.2 | XP_011530658.1 | 22, 23 | 5713 | |||
XM_011532358.2 | XP_011530660.1 | 19, 20 | 1123 | |||
XM_017008735.1 | XP_016864224.1 | 23, 24 | 1630 | |||
XM_017008736.1 | XP_016864225.1 | 23, 24 | 2046 | |||
XM_017008737.1 | XP_016864226.1 | 23, 24 | 4408 | |||
GenBank mRNA | AF293340.1 | - | 24, 25 | 1859 | ||
AF293341.1 | - | 24, 25 | 1859 | |||
BC036669.2 | - | 21, 22 | 1233 | 5268875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
