siRNA Details
Gene Symbol: COL25A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256074.1 | NP_001243003.1 | 26, 27 | 1842 | ||
NM_032518.2 | NP_115907.2 | 29, 30 | 2106 | |||
NM_198721.3 | NP_942014.1 | 29, 30 | 2106 | |||
NR_045756.2 | 26, 27 | 1436 | ||||
XM_011532333.2 | XP_011530635.1 | 29, 30 | 2199 | |||
XM_011532334.2 | XP_011530636.1 | 29, 30 | 2065 | |||
XM_011532335.2 | XP_011530637.1 | 29, 30 | 1894 | |||
XM_011532338.2 | XP_011530640.1 | 27, 28 | 2274 | |||
XM_011532355.2 | XP_011530657.1 | 28, 29 | 5978 | |||
XM_011532356.2 | XP_011530658.1 | 27, 28 | 5960 | |||
XM_011532358.2 | XP_011530660.1 | 24, 25 | 1370 | |||
XM_017008735.1 | XP_016864224.1 | 28, 29 | 1877 | |||
XM_017008736.1 | XP_016864225.1 | 28, 29 | 2293 | |||
XM_017008737.1 | XP_016864226.1 | 28, 29 | 4655 | |||
GenBank mRNA | AF293340.1 | - | 29, 30 | 2106 | ||
AF293341.1 | - | 29, 30 | 2106 | |||
BC036669.2 | - | 26, 27 | 1436 | 5268875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
