siRNA Details
Gene Symbol: COL25A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256074.1 | NP_001243003.1 | 1 | 596 | ||
NM_032518.2 | NP_115907.2 | 2 | 779 | |||
NM_198721.3 | NP_942014.1 | 2 | 779 | |||
NR_045756.2 | 2 | 369 | ||||
XM_011532333.2 | XP_011530635.1 | 2 | 872 | |||
XM_011532334.2 | XP_011530636.1 | 2 | 738 | |||
XM_011532335.2 | XP_011530637.1 | 2 | 567 | |||
XM_011532338.2 | XP_011530640.1 | 1 | 965 | |||
XM_017008735.1 | XP_016864224.1 | 2 | 568 | |||
XM_017008736.1 | XP_016864225.1 | 1 | 966 | |||
GenBank mRNA | AF293340.1 | - | 2 | 779 | ||
AF293341.1 | - | 2 | 779 | |||
BC036669.2 | - | 2 | 369 | 5268875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
