siRNA Details
Gene Symbol: ALG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195223.1 | NP_001182152.1 | 2 | 245 | ||
XM_017005844.1 | XP_016861333.1 | 2 | 3509 | |||
XM_017005845.1 | XP_016861334.1 | 2 | 135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
