siRNA Details
Gene Symbol: OR5D18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001952.1 | NP_001001952.1 | 1 | 658 | ||
GenBank mRNA | BC136925.1 | - | 1 | 678 | 9020917 | |
BC136927.1 | - | 1 | 681 | 9020919 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
