siRNA Details
Gene Symbol: SPDYA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008779.1 | NP_001008779.1 | 5 | 714 | ||
NM_001142634.1 | NP_001136106.1 | 6 | 828 | |||
NM_182756.3 | NP_877433.2 | 7 | 902 | |||
GenBank mRNA | AY820303.1 | - | 5 | 696 | ||
AY820304.1 | - | 5 | 696 | |||
BC093005.1 | - | 6 | 828 | 4839330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
