siRNA Details
Gene Symbol: MIR22HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028502.1 | 2 | 1744 | |||
NR_028503.1 | 3 | 937 | ||||
GenBank mRNA | AF070569.1 | - | 1 | 1313 | ||
BI756450.1 | - | 4 | 848 | |||
BX648321.1 | - | 2 | 1681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
