siRNA Details
Gene Symbol: MIR22HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028502.1 | 2 | 1645 | |||
NR_028503.1 | 3 | 838 | ||||
GenBank mRNA | AF070569.1 | - | 1 | 1214 | ||
BX648321.1 | - | 2 | 1582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
