siRNA Details
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 13 | 1432 | ||
NM_004434.2 | NP_004425.2 | 12 | 1375 | |||
XM_005267397.1 | XP_005267454.1 | 13 | 1502 | |||
XM_005267398.3 | XP_005267455.1 | 13 | 1435 | |||
XM_005267399.4 | XP_005267456.2 | 13 | 1474 | |||
XM_005267400.4 | XP_005267457.1 | 12 | 1401 | |||
XM_011536540.2 | XP_011534842.1 | 12 | 1448 | |||
XM_011536541.1 | XP_011534843.1 | 13 | 1515 | |||
XM_011536542.2 | XP_011534844.1 | 13 | 1474 | |||
XM_017021074.1 | XP_016876563.1 | 12 | 1445 | |||
GenBank mRNA | AK023861.1 | - | 3 | 1064 | ||
AK126980.1 | - | 14 | 1388 | |||
AK299812.1 | - | 13 | 1711 | |||
BC032541.1 | - | 12 | 1351 | 5533599 | ||
BC033043.1 | - | 12 | 1375 | 4821202 | ||
BX247979.1 | - | 13 | 1444 | |||
U97018.1 | - | 12 | 1374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
