siRNA Details
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 8, 9 | 950 | ||
NM_004434.2 | NP_004425.2 | 7, 8 | 893 | |||
XM_005267397.1 | XP_005267454.1 | 8, 9 | 1020 | |||
XM_005267398.3 | XP_005267455.1 | 8, 9 | 953 | |||
XM_005267399.4 | XP_005267456.2 | 8, 9 | 992 | |||
XM_005267400.4 | XP_005267457.1 | 7, 8 | 919 | |||
XM_011536540.2 | XP_011534842.1 | 7, 8 | 966 | |||
XM_011536541.1 | XP_011534843.1 | 8, 9 | 1033 | |||
XM_011536542.2 | XP_011534844.1 | 8, 9 | 992 | |||
XM_017021074.1 | XP_016876563.1 | 7, 8 | 963 | |||
GenBank mRNA | AK126980.1 | - | 9, 10 | 906 | ||
AK299812.1 | - | 8, 9 | 1229 | |||
AK307925.1 | - | 7, 8 | 886 | |||
BC032541.1 | - | 7, 8 | 869 | 5533599 | ||
BC033043.1 | - | 7, 8 | 893 | 4821202 | ||
BX247979.1 | - | 8, 9 | 962 | |||
U97018.1 | - | 7, 8 | 892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
