siRNA Details
Gene Symbol: MPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044370.1 | NP_001037835.1 | 3 | 522 | ||
XM_011530367.2 | XP_011528669.1 | 3 | 4977 | |||
GenBank mRNA | AK289774.1 | - | 3 | 419 | ||
AK290541.1 | - | 3 | 522 | |||
AK294912.1 | - | 3 | 523 | |||
AK295077.1 | - | 3 | 739 | |||
AK297276.1 | - | 4 | 456 | |||
AK299531.1 | - | 2 | 220 | |||
AK316008.1 | - | 3 | 637 | |||
BC028035.1 | - | 2 | 386 | 5242615 | ||
CT841512.1 | - | 2 | 281 | |||
DA773758.1 | - | 2 | 220 | |||
U84894.1 | - | 2 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
