siRNA Details
Gene Symbol: MT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005947.2 | NP_005938.1 | 3 | 215 | ||
GenBank mRNA | AF349000.2 | - | 3 | 214 | ||
AY168638.1 | - | 3 | 216 | |||
BC069421.1 | - | 3 | 210 | 7262703 | ||
BC137478.1 | - | 3 | 182 | 9021478 | ||
BC137479.1 | - | 3 | 182 | 9021479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
