siRNA Details
Gene Symbol: C17orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181655.3 | NP_858041.2 | 3 | 1119 | ||
NM_181656.4 | NP_858042.2 | 3 | 1262 | |||
GenBank mRNA | AK026583.1 | - | 3 | 1200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
