siRNA Details
Gene Symbol: C17orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181656.4 | NP_858042.2 | 2 | 469 | ||
GenBank mRNA | AK026583.1 | - | 2 | 407 | ||
BC021702.1 | - | 1 | 88 | |||
BX364842.2 | - | 2 | 468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
