siRNA Details
Gene Symbol: POFUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015227.4 | NP_056042.1 | 6 | 800 | ||
NM_133635.4 | NP_598368.2 | 6 | 800 | |||
NR_004858.1 | 6 | 800 | ||||
XM_006723981.3 | XP_006724044.1 | 7 | 873 | |||
XM_011529504.2 | XP_011527806.1 | 8 | 1009 | |||
XM_011529505.2 | XP_011527807.1 | 8 | 1010 | |||
XM_011529506.2 | XP_011527808.1 | 7 | 948 | |||
XM_011529507.2 | XP_011527809.1 | 7 | 844 | |||
XM_011529508.2 | XP_011527810.1 | 8 | 1010 | |||
XM_011529510.2 | XP_011527812.1 | 8 | 918 | |||
XM_017028306.1 | XP_016883795.1 | 6 | 783 | |||
XM_017028309.1 | XP_016883798.1 | 4 | 206 | |||
GenBank mRNA | AB023175.1 | - | 6 | 789 | ||
AJ302079.2 | - | 6 | 789 | |||
AJ302080.2 | - | 6 | 789 | |||
AJ575591.1 | - | 6 | 784 | |||
AK290732.1 | - | 6 | 797 | |||
AL110285.1 | - | 6 | 786 | |||
AY066015.2 | - | 6 | 789 | |||
BC011044.1 | - | 2 | 248 | |||
BC064623.1 | - | 6 | 782 | 5727996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
