siRNA Details
Gene Symbol: CA5BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026551.1 | 5 | 572 | |||
GenBank mRNA | BC032506.1 | - | 1 | 775 | ||
BQ228866.1 | - | 1 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
