siRNA Details
Gene Symbol: CA5BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026551.1 | 5 | 746 | |||
GenBank mRNA | BC021816.1 | - | 3 | 174 | ||
BC032506.1 | - | 1 | 949 | |||
BQ228866.1 | - | 1 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
