siRNA Details
Gene Symbol: FAM91A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317917.1 | NP_001304846.1 | 1, 2 | 342 | ||
NM_001317918.1 | NP_001304847.1 | 1, 2 | 342 | |||
NM_144963.3 | NP_659400.2 | 1, 2 | 342 | |||
GenBank mRNA | AK300603.1 | - | 1, 2 | 325 | ||
BC030520.1 | - | 1, 2 | 318 | |||
BC144630.1 | - | 1, 2 | 135 | |||
CN263780.1 | - | 1, 2 | 331 | |||
DC308945.1 | - | 1, 2 | 342 | |||
DQ228141.1 | - | 1, 2 | 139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
