siRNA Details
Gene Symbol: PRELID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138492.5 | NP_612501.3 | 5 | 613 | ||
NM_182960.3 | NP_892005.1 | 5 | 446 | |||
NM_205846.2 | NP_995318.1 | 4 | 410 | |||
XM_017009127.1 | XP_016864616.1 | 5 | 446 | |||
XM_017009128.1 | XP_016864617.1 | 5 | 446 | |||
XM_017009129.1 | XP_016864618.1 | 5 | 446 | |||
XM_017009130.1 | XP_016864619.1 | 5 | 446 | |||
XM_017009131.1 | XP_016864620.1 | 5 | 446 | |||
XM_017009132.1 | XP_016864621.1 | 5 | 446 | |||
XM_017009133.1 | XP_016864622.1 | 5 | 446 | |||
XM_017009134.1 | XP_016864623.1 | 5 | 446 | |||
XM_017009135.1 | XP_016864624.1 | 4 | 410 | |||
GenBank mRNA | AK095695.1 | - | 5 | 381 | ||
AK125215.1 | - | 4 | 345 | |||
BC012033.1 | - | 5 | 580 | 4476359 | ||
BC028175.1 | - | 327 | ||||
BQ645893.1 | - | 4 | 380 | |||
BU167299.1 | - | 4 | 272 | |||
BU186346.1 | - | 5 | 383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
