siRNA Details
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127893.1 | NP_001121365.1 | 8 | 2175 | ||
NM_020219.3 | NP_064604.2 | 8 | 2178 | |||
GenBank mRNA | AK128234.1 | - | 4 | 3202 | ||
AL390165.1 | - | 1 | 561 | |||
BC083499.1 | - | 8 | 2175 | 30390224 | ||
KJ851726.1 | - | 8 | 1723 | |||
KJ851727.1 | - | 8 | 1726 | |||
KJ851728.1 | - | 9 | 1758 | |||
KJ851729.1 | - | 9 | 1761 | |||
KJ851730.1 | - | 9 | 1762 | |||
KJ851731.1 | - | 9 | 1765 | |||
KJ851732.1 | - | 7 | 1611 | |||
KJ851733.1 | - | 7 | 1614 | |||
KJ851734.1 | - | 7 | 1639 | |||
KJ851735.1 | - | 7 | 1642 | |||
KJ851736.1 | - | 8 | 1674 | |||
KJ851737.1 | - | 8 | 1677 | |||
KJ851738.1 | - | 8 | 1678 | |||
KJ851739.1 | - | 8 | 1681 | |||
KJ851740.1 | - | 5 | 1478 | |||
KJ851741.1 | - | 5 | 1481 | |||
KJ851742.1 | - | 7 | 1544 | |||
KJ851743.1 | - | 7 | 1547 | |||
KJ851744.1 | - | 7 | 1572 | |||
KJ851745.1 | - | 7 | 1575 | |||
KJ851746.1 | - | 8 | 1607 | |||
KJ851747.1 | - | 8 | 1610 | |||
KJ851748.1 | - | 8 | 1611 | |||
KJ851749.1 | - | 8 | 1614 | |||
KJ851750.1 | - | 6 | 1460 | |||
KJ851751.1 | - | 6 | 1463 | |||
KJ851752.1 | - | 6 | 1488 | |||
KJ851753.1 | - | 6 | 1491 | |||
KJ851754.1 | - | 7 | 1523 | |||
KJ851755.1 | - | 7 | 1526 | |||
KJ851756.1 | - | 7 | 1527 | |||
KJ851757.1 | - | 7 | 1530 | |||
KJ851758.1 | - | 4 | 1327 | |||
KJ851759.1 | - | 4 | 1330 | |||
KJ851760.1 | - | 9 | 1829 | |||
KJ851761.1 | - | 9 | 1832 | |||
KJ851762.1 | - | 9 | 1857 | |||
KJ851763.1 | - | 9 | 1860 | |||
KJ851764.1 | - | 10 | 1892 | |||
KJ851765.1 | - | 10 | 1895 | |||
KJ851766.1 | - | 11 | 1896 | |||
KJ851767.1 | - | 11 | 1899 | |||
KJ851768.1 | - | 8 | 1745 | |||
KJ851769.1 | - | 8 | 1748 | |||
KJ851770.1 | - | 8 | 1773 | |||
KJ851771.1 | - | 8 | 1776 | |||
KJ851772.1 | - | 9 | 1808 | |||
KJ851773.1 | - | 9 | 1811 | |||
KJ851774.1 | - | 9 | 1812 | |||
KJ851775.1 | - | 9 | 1815 | |||
KJ851776.1 | - | 6 | 1612 | |||
KJ851777.1 | - | 6 | 1615 | |||
KJ851778.1 | - | 8 | 1678 | |||
KJ851779.1 | - | 8 | 1681 | |||
KJ851780.1 | - | 8 | 1706 | |||
KJ851781.1 | - | 8 | 1709 | |||
KJ851782.1 | - | 9 | 1741 | |||
KJ851783.1 | - | 9 | 1744 | |||
KJ851784.1 | - | 9 | 1745 | |||
KJ851785.1 | - | 9 | 1748 | |||
KJ851786.1 | - | 7 | 1594 | |||
KJ851787.1 | - | 7 | 1597 | |||
KJ851788.1 | - | 7 | 1622 | |||
KJ851789.1 | - | 7 | 1625 | |||
KJ851790.1 | - | 8 | 1657 | |||
KJ851791.1 | - | 8 | 1660 | |||
KJ851792.1 | - | 8 | 1661 | |||
KJ851793.1 | - | 8 | 1664 | |||
KJ851794.1 | - | 5 | 1461 | |||
KJ851795.1 | - | 5 | 1464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
