siRNA Details
Gene Symbol: KRTAP10-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198690.2 | NP_941963.2 | 1 | 776 | ||
GenBank mRNA | AB076356.1 | - | 1 | 776 | ||
BC131613.1 | - | 1 | 752 | 40115414 |
Gene Symbol: KRTAP10-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198687.2 | NP_941960.2 | 1 | 1147 | ||
GenBank mRNA | AB076351.1 | - | 1 | 1147 | ||
BC125048.1 | - | 1 | 1136 | 40116133 | ||
BC125049.1 | - | 1 | 1136 | 40116140 |
Gene Symbol: KRTAP10-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB076352.1 | - | 1 | 723 | ||
BC120949.1 | - | 1 | 722 | 40115143 | ||
BC120950.1 | - | 1 | 722 | 40115146 |
Gene Symbol: KRTAP10-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198689.2 | NP_941962.1 | 1 | 986 | ||
GenBank mRNA | AB076354.1 | - | 1 | 986 | ||
AJ566385.1 | - | 1 | 12 | |||
BC119643.2 | - | 1 | 983 | 40115724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
