siRNA Details
Gene Symbol: RASSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032023.3 | NP_114412.2 | 9 | 887 | ||
XM_005271833.4 | XP_005271890.1 | 9 | 937 | |||
XM_005271835.4 | XP_005271892.1 | 9 | 887 | |||
XM_006718013.3 | XP_006718076.1 | 9 | 982 | |||
XM_006718014.3 | XP_006718077.1 | 8 | 2401 | |||
XM_011540255.2 | XP_011538557.1 | 8 | 762 | |||
XM_017016754.1 | XP_016872243.1 | 8 | 2527 | |||
XM_017016755.1 | XP_016872244.1 | 8 | 2482 | |||
XM_017016756.1 | XP_016872245.1 | 9 | 1008 | |||
XM_017016757.1 | XP_016872246.1 | 8 | 2553 | |||
XM_017016758.1 | XP_016872247.1 | 7 | 2258 | |||
GenBank mRNA | AB209446.1 | - | 8 | 2759 | ||
AF260335.1 | - | 9 | 851 | |||
AK055763.1 | - | 7 | 1895 | |||
AK092770.1 | - | 6 | 2098 | |||
AK097272.1 | - | 4 | 2165 | |||
AL713716.1 | - | 1 | 60 | |||
AY216713.1 | - | 9 | 887 | |||
AY216714.1 | - | 9 | 847 | |||
AY216715.1 | - | 9 | 914 | |||
AY216716.1 | - | 8 | 795 | |||
BC032593.1 | - | 9 | 869 | 5527647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
