siRNA Details
Gene Symbol: RASSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032023.3 | NP_114412.2 | 8 | 808 | ||
XM_005271833.4 | XP_005271890.1 | 8 | 858 | |||
XM_005271835.4 | XP_005271892.1 | 8 | 808 | |||
XM_006718013.3 | XP_006718076.1 | 8 | 903 | |||
XM_006718014.3 | XP_006718077.1 | 7 | 2322 | |||
XM_011540255.2 | XP_011538557.1 | 7 | 683 | |||
XM_017016754.1 | XP_016872243.1 | 7 | 2448 | |||
XM_017016755.1 | XP_016872244.1 | 7 | 2403 | |||
XM_017016756.1 | XP_016872245.1 | 8 | 929 | |||
XM_017016757.1 | XP_016872246.1 | 7 | 2474 | |||
XM_017016758.1 | XP_016872247.1 | 6 | 2179 | |||
GenBank mRNA | AB209446.1 | - | 7 | 2680 | ||
AF260335.1 | - | 8 | 772 | |||
AK055763.1 | - | 6 | 1816 | |||
AK092770.1 | - | 5 | 2018 | |||
AK097272.1 | - | 3 | 2086 | |||
AY216713.1 | - | 8 | 808 | |||
AY216714.1 | - | 8 | 768 | |||
AY216715.1 | - | 8 | 835 | |||
AY216716.1 | - | 7 | 716 | |||
BC032593.1 | - | 8 | 790 | 5527647 | ||
BI601296.1 | - | 8 | 809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
