siRNA Details
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 2 | 427 | |||
NR_026926.1 | 2 | 427 | ||||
GenBank mRNA | AF451986.1 | - | 1 | 18 | ||
AK057399.1 | - | 2 | 363 | |||
DA518680.1 | - | 2 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
