siRNA Details
Gene Symbol: SUPT3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261823.1 | NP_001248752.1 | 1 | 266 | ||
NM_003599.3 | NP_003590.1 | 1 | 266 | |||
NM_181356.2 | NP_852001.1 | 1 | 266 | |||
XM_011514950.1 | XP_011513252.1 | 1 | 266 | |||
GenBank mRNA | AF073930.1 | - | 1 | 53 | ||
AK303838.1 | - | 1 | 79 | |||
AK313874.1 | - | 1 | 256 | |||
BC035816.1 | - | 1 | 143 | |||
BC050384.1 | - | 1 | 148 | 5758833 | ||
DA012115.1 | - | 1 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
