siRNA Details
Gene Symbol: SUPT3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261823.1 | NP_001248752.1 | 2 | 374 | ||
NM_003599.3 | NP_003590.1 | 2 | 374 | |||
NM_181356.2 | NP_852001.1 | 2 | 374 | |||
XM_011514949.2 | XP_011513251.1 | 2 | 210 | |||
XM_011514950.1 | XP_011513252.1 | 2 | 374 | |||
XM_011514951.2 | XP_011513253.1 | 2 | 4435 | |||
XM_011514952.2 | XP_011513254.1 | 2 | 1421 | |||
XM_011514953.2 | XP_011513255.1 | 2 | 204 | |||
XM_011514954.2 | XP_011513256.1 | 2 | 235 | |||
XM_017011369.1 | XP_016866858.1 | 2 | 4438 | |||
XM_017011371.1 | XP_016866860.1 | 3 | 3549 | |||
XM_017011372.1 | XP_016866861.1 | 2 | 263 | |||
XM_017011373.1 | XP_016866862.1 | 2 | 227 | |||
GenBank mRNA | AF069734.1 | - | 1 | 90 | ||
AF073930.1 | - | 2 | 161 | |||
AK303838.1 | - | 2 | 187 | |||
AK313874.1 | - | 2 | 364 | |||
BC035816.1 | - | 2 | 251 | |||
BC050384.1 | - | 2 | 256 | 5758833 | ||
DA012115.1 | - | 2 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
