siRNA Details
Gene Symbol: SUPT3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181356.2 | NP_852001.1 | 4 | 511 | ||
XM_011514949.2 | XP_011513251.1 | 4 | 347 | |||
XM_017011369.1 | XP_016866858.1 | 4 | 4575 | |||
XM_017011370.1 | XP_016866859.1 | 2 | 140 | |||
GenBank mRNA | BC050384.1 | - | 4 | 393 | 5758833 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
