siRNA Details
Gene Symbol: CEP104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014704.3 | NP_055519.1 | 4 | 728 | ||
XM_005244815.4 | XP_005244872.1 | 5 | 828 | |||
XM_011542473.2 | XP_011540775.1 | 5 | 1028 | |||
XM_011542474.2 | XP_011540776.1 | 4 | 746 | |||
XM_011542475.2 | XP_011540777.1 | 5 | 1028 | |||
XM_011542476.2 | XP_011540778.1 | 5 | 1028 | |||
XM_011542477.2 | XP_011540779.1 | 5 | 1028 | |||
XM_011542478.1 | XP_011540780.1 | 5 | 1028 | |||
XM_017002917.1 | XP_016858406.1 | 5 | 1010 | |||
XM_017002918.1 | XP_016858407.1 | 4 | 728 | |||
XM_017002919.1 | XP_016858408.1 | 4 | 706 | |||
GenBank mRNA | AB011134.1 | - | 4 | 682 | ||
AK308191.1 | - | 4 | 687 | |||
BC001640.2 | - | 4 | 693 | |||
BC047450.1 | - | 4 | 693 | 5297035 | ||
BC050721.1 | - | 4 | 709 | |||
DA081362.1 | - | 2 | 170 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
