siRNA Details
Gene Symbol: CEP104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014704.3 | NP_055519.1 | 4 | 757 | ||
XM_005244815.4 | XP_005244872.1 | 5 | 857 | |||
XM_011542473.2 | XP_011540775.1 | 5 | 1057 | |||
XM_011542474.2 | XP_011540776.1 | 4 | 775 | |||
XM_011542475.2 | XP_011540777.1 | 5 | 1057 | |||
XM_011542476.2 | XP_011540778.1 | 5 | 1057 | |||
XM_011542477.2 | XP_011540779.1 | 5 | 1057 | |||
XM_011542478.1 | XP_011540780.1 | 5 | 1057 | |||
XM_017002917.1 | XP_016858406.1 | 5 | 1039 | |||
XM_017002918.1 | XP_016858407.1 | 4 | 757 | |||
XM_017002919.1 | XP_016858408.1 | 4 | 735 | |||
GenBank mRNA | AB011134.1 | - | 4 | 711 | ||
AK308191.1 | - | 4 | 716 | |||
BC001640.2 | - | 4 | 722 | |||
BC047450.1 | - | 4 | 722 | 5297035 | ||
BC050721.1 | - | 4 | 738 | |||
DA081362.1 | - | 2 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
