siRNA Details
Gene Symbol: SF3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012433.3 | NP_036565.2 | 12 | 1730 | ||
GenBank mRNA | AF054284.1 | - | 12 | 1636 | ||
AK308122.1 | - | 12 | 1682 | |||
BC039020.1 | - | 12 | 1653 | |||
BC056155.1 | - | 12 | 1680 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_031179, AB037890, AK140982.
Gene Ontology Categories:
