siRNA Details
Gene Symbol: SF3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012433.3 | NP_036565.2 | 10 | 1373 | ||
GenBank mRNA | AF054284.1 | - | 10 | 1279 | ||
AK308122.1 | - | 10 | 1325 | |||
BC015530.1 | - | 10 | 1310 | |||
BC029418.1 | - | 10 | 1309 | |||
BC039020.1 | - | 10 | 1296 | |||
BC056155.1 | - | 10 | 1323 | |||
BC064335.1 | - | 10 | 1294 | |||
BC107883.1 | - | 10 | 1300 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_031179, AK146989, BC060125, BC003419, BC080754, and others.
Gene Ontology Categories:
