siRNA Details
Gene Symbol: SF3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012433.3 | NP_036565.2 | 25 | 3944 | ||
GenBank mRNA | AF054284.1 | - | 25 | 3850 | ||
AF070540.1 | - | 5 | 822 | |||
AL137410.1 | - | 3 | 367 | |||
CD692669.1 | - | 4 | 482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
