siRNA Details
Gene Symbol: ZNF436-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033690.1 | 1 | 2727 | |||
NR_033691.1 | 3 | 552 | ||||
GenBank mRNA | AK074989.1 | - | 1 | 2642 | ||
BC028411.1 | - | 1 | 2468 | 4822173 | ||
BC054115.1 | - | 1 | 2731 | 5270027 | ||
BC068594.1 | - | 1 | 2709 | 4828713 | ||
BX098506.1 | - | 3 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
