siRNA Details
Gene Symbol: RPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001026.4 | NP_001017.1 | 2 | 159 | ||
NM_001142282.1 | NP_001135754.1 | 2 | 159 | |||
NM_001142283.1 | NP_001135755.1 | 2 | 159 | |||
NM_001142284.1 | NP_001135756.1 | 2 | 159 | |||
NM_001142285.1 | NP_001135757.1 | 2 | 159 | |||
NM_033022.3 | NP_148982.1 | 2 | 159 | |||
GenBank mRNA | AF147330.1 | - | 2 | 40 | ||
AK311776.1 | - | 2 | 56 | |||
AK311910.1 | - | 2 | 56 | |||
BC000523.2 | - | 2 | 36 | 2961125 | ||
BC003149.1 | - | 2 | 32 | |||
BC071926.1 | - | 2 | 41 | 5556377 | ||
BG610671.1 | - | 103 | ||||
BG944046.1 | - | 2 | 160 | |||
BM745909.1 | - | 2 | 56 | |||
BX537975.1 | - | 2 | 55 | |||
BX647260.1 | - | 2 | 45 | |||
CD176901.1 | - | 2 | 82 | |||
CD677556.1 | - | 2 | 35 | |||
DB116225.1 | - | 2 | 56 | |||
M31520.1 | - | 2 | 159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
