siRNA Details
Gene Symbol: H2AFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040158.1 | NP_001035248.1 | 4 | 659 | ||
NM_004893.2 | NP_004884.1 | 4 | 622 | |||
NM_138609.2 | NP_613075.1 | 4 | 659 | |||
NM_138610.2 | NP_613258.2 | 4 | 659 | |||
XM_005272132.1 | XP_005272189.1 | 4 | 1037 | |||
XM_005272134.3 | XP_005272191.1 | 4 | 1037 | |||
XM_005272135.3 | XP_005272192.1 | 4 | 1037 | |||
XM_011543729.2 | XP_011542031.1 | 4 | 576 | |||
XM_011543730.2 | XP_011542032.1 | 4 | 579 | |||
XM_011543731.1 | XP_011542033.1 | 4 | 1037 | |||
XM_011543732.1 | XP_011542034.1 | 4 | 1037 | |||
XM_011543733.2 | XP_011542035.1 | 4 | 1037 | |||
XM_011543734.2 | XP_011542036.1 | 4 | 1037 | |||
XM_011543735.2 | XP_011542037.1 | 4 | 1037 | |||
XM_017010067.1 | XP_016865556.1 | 4 | 1037 | |||
XM_017010068.1 | XP_016865557.1 | 4 | 1037 | |||
XM_017010069.1 | XP_016865558.1 | 4 | 1037 | |||
GenBank mRNA | AF041483.1 | - | 4 | 556 | ||
AF044286.1 | - | 3 | 451 | |||
AF054174.1 | - | 4 | 622 | |||
AK023409.1 | - | 4 | 650 | |||
AY134746.1 | - | 3 | 435 | |||
BC013331.1 | - | 4 | 585 | 4077577 | ||
BC095406.1 | - | 4 | 619 | 30348703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
